Learn More
Chemglass Life Sciences 3000mL Single Neck Round Bottom Flask (RBF) , 35/20 Socket Joint
SDP
Catalog No. CG150836
Encompass_Preferred
3000mL Single Neck Round Bottomed Flask, 35/20 Socket Joint. Round bottom, HEAVY WALL single short neck flask with a spherical socket joint. Flasks having a 35/20 socket joint are used as replacements for the collection flask on rotary evaporators.
Catalog No. CG150836 Supplier Chemglass Life Sciences Supplier No. CG150836
3000mL Single Neck Round Bottomed Flask, 35/20 Socket Joint. Round bottom, HEAVY WALL single short neck flask with a spherical socket joint. Flasks having a 35/20 socket joint are used as replacements for the collection flask on rotary evaporators.