Learn More
Chemglass Life Sciences 3000mL Reaction Vessel, Cylindrical, Jacketed, 150mm Schott O-Ring Flange, Approx body OD x Height 143mm I.D. x 275mm Height, Hose Conn. Thread Size GL-14
SDP
Catalog No. CG192604
Encompass_Preferred
3000mL Reaction Vessel, Cylindrical, Jacketed, 150mm Schott O-Ring Flange, 143mm I.D. x 275mm Height, Hose Conn. Thread Size GL-14. Cylindrical, jacketed, reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Cooling jacket is equipped with two removable hose connections to reduce the risk of injury while attaching or detaching flexible tubing. Supplied complete with two polypropylene hose connections and two screw caps of the listed size. For clamp see CG-141 or CG-141-T. For o-ring see CG-147.PLEASE NOTE: HIGH TEMPERATURE HOSE CONNECTIONS AND SCREW CAPS ARE AVAILABLE. SEE CG-195-H AND CG-198-H.
Catalog No. CG192604 Supplier Chemglass Life Sciences Supplier No. CG192604
3000mL Reaction Vessel, Cylindrical, Jacketed, 150mm Schott O-Ring Flange, 143mm I.D. x 275mm Height, Hose Conn. Thread Size GL-14. Cylindrical, jacketed, reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Cooling jacket is equipped with two removable hose connections to reduce the risk of injury while attaching or detaching flexible tubing. Supplied complete with two polypropylene hose connections and two screw caps of the listed size. For clamp see CG-141 or CG-141-T. For o-ring see CG-147.PLEASE NOTE: HIGH TEMPERATURE HOSE CONNECTIONS AND SCREW CAPS ARE AVAILABLE. SEE CG-195-H AND CG-198-H.