Learn More
Chemglass Life Sciences 3L Reaction Vessel, Jacketed, 150mm Schott O-Ring Flange, 3/4" Beaded Pipe Inlet/Outlet, 1-1/2" Beaded Pipe Detachable Drain Valve
SDP
Catalog No. 501531276
Encompass_Preferred
3L Reaction Vessel, Jacketed, 150mm Schott O-Ring Flange, 3/4" Beaded Pipe Inlet/Outlet, 1-1/2" Beaded Pipe Detachable Drain Valve. Jacketed reaction vessel with graduation scale on both sides. Vessels are available with Chemglass's 1-1/2 beaded pipe detachable drain valve. Reaction vessel is constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Inlet/outlet is a 3/4 beaded pipe fitting. For clamp see CG-141 or CG-141-T. For o-ring see CG-147.PLEASE NOTE: VALVE, BEADED PIPE COUPLINGS, AND ADAPTERS MUST BE PURCHASED SEPARATELY.
Catalog No. 50-153-1276 Supplier Chemglass Life Sciences Supplier No. CG192923
3L Reaction Vessel, Jacketed, 150mm Schott O-Ring Flange, 3/4" Beaded Pipe Inlet/Outlet, 1-1/2" Beaded Pipe Detachable Drain Valve. Jacketed reaction vessel with graduation scale on both sides. Vessels are available with Chemglass's 1-1/2 beaded pipe detachable drain valve. Reaction vessel is constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Inlet/outlet is a 3/4 beaded pipe fitting. For clamp see CG-141 or CG-141-T. For o-ring see CG-147.PLEASE NOTE: VALVE, BEADED PIPE COUPLINGS, AND ADAPTERS MUST BE PURCHASED SEPARATELY.