Learn More
Chemglass Life Sciences Chemglass 3,000 mL Single-Neck Heavy-Wall Wide-Mouth Recovery Flask, GL80, Complete (CG-1512-X-03)
SDP
Catalog No. NC2190865
Encompass_Preferred
3,000 mL single-neck heavy-wall wide-mouth recovery flask with a GL80 glass thread, supplied complete with a PTFE GL80-to-24/40 adapter. The heavy-wall construction is suitable for vacuum applications and the balanced contour minimizes run-out during rotary evaporation. The wide mouth allows easy emptying and cleaning, and the PTFE adapter uses an FDA-grade silicone gasket so liquids or powders contact only glass or PTFE.
  • Capacity: 3,000 mL.
  • Thread size: GL80 wide-mouth glass thread.
  • Includes: PTFE GL80-to-24/40 adapter with FDA-grade silicone gasket.
  • Construction: Heavy-wall glass suitable for vacuum applications.
  • Design: Balanced contour to prevent run-out during rotation.
  • Cleaning and handling: Wide mouth for easy emptying and access with tools.
  • Typical use: Recovery and evaporative processes in rotary evaporators.
This item is not returnable. View return policy
Catalog No. NC2190865 Supplier Chemglass Life Sciences Supplier No. CG1512X03
3,000 mL single-neck heavy-wall wide-mouth recovery flask with a GL80 glass thread, supplied complete with a PTFE GL80-to-24/40 adapter. The heavy-wall construction is suitable for vacuum applications and the balanced contour minimizes run-out during rotary evaporation. The wide mouth allows easy emptying and cleaning, and the PTFE adapter uses an FDA-grade silicone gasket so liquids or powders contact only glass or PTFE.
  • Capacity: 3,000 mL.
  • Thread size: GL80 wide-mouth glass thread.
  • Includes: PTFE GL80-to-24/40 adapter with FDA-grade silicone gasket.
  • Construction: Heavy-wall glass suitable for vacuum applications.
  • Design: Balanced contour to prevent run-out during rotation.
  • Cleaning and handling: Wide mouth for easy emptying and access with tools.
  • Typical use: Recovery and evaporative processes in rotary evaporators.