Learn More
Ace Glass, Inc. Fabric heating mantle that covers top and bottom of 1, 2 or3 neck 3000mL spherical flask. Lower half wattage is 500w at115 volts. Upper half wattage is 200w at 115 volts d, .
SDP
Catalog No. 50977261
Encompass
Catalog No. 50-977-261 Supplier Ace Glass, Inc. Supplier No. 1203123