Learn More
Chemglass Life Sciences Stirrer Blade, PTFE, 19mm, Heavy Duty, for 3000mL Flasks, 35mm Width x 93mm Length, 45/50 minimum joint clearance
SDP
Catalog No. 50872515
Encompass_Preferred
Stirrer Blade, PTFE, 19mm, Heavy Duty, for 3000mL Flasks, 35mm Width x 93mm Length, 45/50 minimum joint clearance. Stirrer blade made of 3/16" thick solid PTFE for use with both Chem-Stir™ and button style stirring shafts having a 19mm OD.
Catalog No. 50-872-515 Supplier Chemglass Life Sciences Supplier No. CG208902
Stirrer Blade, PTFE, 19mm, Heavy Duty, for 3000mL Flasks, 35mm Width x 93mm Length, 45/50 minimum joint clearance. Stirrer blade made of 3/16" thick solid PTFE for use with both Chem-Stir™ and button style stirring shafts having a 19mm OD.