Learn More
Ace Glass, Inc. 3000mL vacuum trap 180 x 410mm w/#25 thread
SDP
Catalog No. 50976309
Encompass_Preferred
3000mL vacuum trap with #25 Ace-Thred includes round bottom flask, adjustable inner tube, and nylon bushing with FETFE o-ring. Flask dimensions: 180mm x 410mm.
Catalog No. 50-976-309 Supplier Ace Glass, Inc. Supplier No. 874425
3000mL vacuum trap with #25 Ace-Thred includes round bottom flask, adjustable inner tube, and nylon bushing with FETFE o-ring. Flask dimensions: 180mm x 410mm.