Learn More
Bellbrook Labs Transcreener GDP FP Assay, Competitive Fluorescence Polarization Immunoassay, 10,000 assays, Far-red fluorescence, Homogeneous, mix-and-read format
SDP
Catalog No. NC1990320
Encompass
Transcreener GDP FP Assay, Competitive Fluorescence Polarization Immunoassay, 10,000 assays, Far-red fluorescence, Homogeneous, mix-and-read format
Catalog No. NC1990320 Supplier Bellbrook Labs Supplier No. 300910K
Transcreener GDP FP Assay, Competitive Fluorescence Polarization Immunoassay, 10,000 assays, Far-red fluorescence, Homogeneous, mix-and-read format