Learn More
CELLTREAT Scientific Products 300cm2 Tissue Culture Flask - Plug Seal Cap, Sterile
SDP
Catalog No. NC2072446
Encompass_Preferred
Tissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachment. Stable base enables filling with flask standing upright. 182cm2 flask offers more growth area than standard 175cm2 flasks. 300cm2 flask offers a large culture area with the convenience of a single layer flask. Convenient zip-closure bag is a practical alternative to folding and taping opened bagsVent caps feature 0.22μm hydrophobic membrane. Large opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapers. Molded, interlocking T design for improved stacking - included on 75, 182 & 300cm² and 250, 600 & 850mL size flasks only. Flask is printed with lot number and expiration date for easy identification and tracking. Angled design is easier to access than square or U-shaped flasks.
Catalog No. NC2072446 Supplier CELLTREAT Scientific Products Supplier No. 229360
Tissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachment. Stable base enables filling with flask standing upright. 182cm2 flask offers more growth area than standard 175cm2 flasks. 300cm2 flask offers a large culture area with the convenience of a single layer flask. Convenient zip-closure bag is a practical alternative to folding and taping opened bagsVent caps feature 0.22μm hydrophobic membrane. Large opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapers. Molded, interlocking T design for improved stacking - included on 75, 182 & 300cm² and 250, 600 & 850mL size flasks only. Flask is printed with lot number and expiration date for easy identification and tracking. Angled design is easier to access than square or U-shaped flasks.