Learn More
Ace Glass, Inc. 300mL 55/50 extraction apparatus
SDP
Catalog No. 50978873
Encompass_Preferred
300ml Soxhlet extraction apparatus with 55/50 joint, uses Friedrichs Condenser in place of Allihn Condenser. Apparatus consists of flask cap, condenser, extractor, and flask.
Catalog No. 50-978-873 Supplier Ace Glass, Inc. Supplier No. 671829
300ml Soxhlet extraction apparatus with 55/50 joint, uses Friedrichs Condenser in place of Allihn Condenser. Apparatus consists of flask cap, condenser, extractor, and flask.