Learn More
Ace Glass, Inc. Erlenmeyer condenser flask, 300mL, #25 Ace-Thred™; neck
SDP
Catalog No. 50975891
Encompass_Preferred
300mL Erlenmeyer condenser flask with a #25 Ace-Thred nylon bushing for connection to the condenser tube. Used with 2000mL reaction kettle with sublimation apparatus accessories.
Catalog No. 50-975-891 Supplier Ace Glass, Inc. Supplier No. 802705
300mL Erlenmeyer condenser flask with a #25 Ace-Thred nylon bushing for connection to the condenser tube. Used with 2000mL reaction kettle with sublimation apparatus accessories.