Learn More
Ace Glass, Inc. 300ML DIST APPR C 300Ml Dist Appr C
SDP
Catalog No. 50973250
Encompass_Preferred
Complete distillation apparatus generally used for the determination of ammonia in water. Joints are standard taper 24/40.
Catalog No. 50-973-250 Supplier Ace Glass, Inc. Supplier No. 655305
Complete distillation apparatus generally used for the determination of ammonia in water. Joints are standard taper 24/40.