Learn More
Chemglass Life Sciences 300mL Erlenmeyer Flask, 24/40 Outer Joint, Graduated
SDP
Catalog No. CG154207
Encompass_Preferred
300mL Erlenmeyer Flask, 24/40 Outer Joint, Graduated. Erlenmeyer flask with a standard taper outer joint. For stoppers see CG-3000.
Catalog No. CG154207 Supplier Chemglass Life Sciences Supplier No. CG154207
300mL Erlenmeyer Flask, 24/40 Outer Joint, Graduated. Erlenmeyer flask with a standard taper outer joint. For stoppers see CG-3000.