Learn More
Ace Glass, Inc. Combo mantle, 6 position, low temperature, fits 250 to 300mLflat bottom flasks, max diameter 3.41 inches, 120v 125 watts per position, order code (-65) or (-75) power supply separately
SDP
Catalog No. 50977350
Encompass
Combo mantle, 6 position, low temperature, fits 250 to 300mLflat bottom flasks, max diameter 3.41 inches, 120v 125 watts per position, order code (-65) or (-75) power supply separately
Catalog No. 50-977-350 Supplier Ace Glass, Inc. Supplier No. 1206111
Combo mantle, 6 position, low temperature, fits 250 to 300mLflat bottom flasks, max diameter 3.41 inches, 120v 125 watts per position, order code (-65) or (-75) power supply separately