Learn More
Ace Glass, Inc. Combo mantle, 6 position, high temperature, fits 250 to 300mLflat bottom flasks, max diameter 3.41 inches, 240v 210 watts per position, order code (-67) or (-77) power supply separately
SDP
Catalog No. 50977353
Encompass
Combo mantle, 6 position, high temperature, fits 250 to 300mLflat bottom flasks, max diameter 3.41 inches, 240v 210 watts per position, order code (-67) or (-77) power supply separately
Catalog No. 50-977-353 Supplier Ace Glass, Inc. Supplier No. 1206129
Combo mantle, 6 position, high temperature, fits 250 to 300mLflat bottom flasks, max diameter 3.41 inches, 240v 210 watts per position, order code (-67) or (-77) power supply separately