Learn More
Ace Glass, Inc. Combo mantle, 6 position, low temperature, fits 250 to 300mLflat bottom flasks, max diameter 3.41 inches, 240v 125 watts per position, order code (-67) or (-77) power supply separately
SDP
Catalog No. 50983289
Encompass
Combo mantle, 6 position, low temperature, fits 250 to 300mLflat bottom flasks, max diameter 3.41 inches, 240v 125 watts per position, order code (-67) or (-77) power supply separately
Catalog No. 50-983-289 Supplier Ace Glass, Inc. Supplier No. 1206119
Combo mantle, 6 position, low temperature, fits 250 to 300mLflat bottom flasks, max diameter 3.41 inches, 240v 125 watts per position, order code (-67) or (-77) power supply separately