Learn More
Ace Glass, Inc. Flask, freeze drying, 300mL, 156mm height, 53mm ID neck, 60mm body OD, includes snap-on rubber cap and 3/4in ODPTFE valved manifold connection adapter
SDP
Catalog No. 5097430
Encompass_Preferred
Catalog No. 50-974-30 Supplier Ace Glass, Inc. Supplier No. 703536