Learn More
Chemglass Life Sciences 300mL Reaction Vessel, Jacketed, 100mm Schott O-Ring Flange, 3/4" Beaded Pipe Inlet/Outlet, CG-911-A-10 Drain Valve
SDP
Catalog No. 501531274
Encompass_Preferred
300mL Reaction Vessel, Jacketed, 100mm Schott O-Ring Flange, 3/4" Beaded Pipe Inlet/Outlet, CG-911-A-10 Drain Valve. Jacketed reaction vessel with graduation scale on both sides. Supplied complete with Chemglass 0-14mm drain valve, one-piece plug, and control knob with red indicator band used for high temperature applications up to 230 C. Siidearm on the CG-911-A valve is 1/2in Swagelok. Vessels with capacities 2L and larger, use the CG-511-02 style drain valve. The sidearm on the CG-511-02 valve is 3/4in Swagelok. Both valves are designed to seal flush with the bottom of the flask avoiding the dead space normally associated with stopcock drains. This permits total mixing and draining of the contents of the reactor. Reaction vessel is constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Inlet/outlet is a 3/4in beaded pipe fitting.
Catalog No. 50-153-1274 Supplier Chemglass Life Sciences Supplier No. CG192911
300mL Reaction Vessel, Jacketed, 100mm Schott O-Ring Flange, 3/4" Beaded Pipe Inlet/Outlet, CG-911-A-10 Drain Valve. Jacketed reaction vessel with graduation scale on both sides. Supplied complete with Chemglass 0-14mm drain valve, one-piece plug, and control knob with red indicator band used for high temperature applications up to 230 C. Siidearm on the CG-911-A valve is 1/2in Swagelok. Vessels with capacities 2L and larger, use the CG-511-02 style drain valve. The sidearm on the CG-511-02 valve is 3/4in Swagelok. Both valves are designed to seal flush with the bottom of the flask avoiding the dead space normally associated with stopcock drains. This permits total mixing and draining of the contents of the reactor. Reaction vessel is constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Inlet/outlet is a 3/4in beaded pipe fitting.