Learn More
Integra Biosciences Corp 300 ml Automation Friendly Reagent Reservoir - Polypropylene | 30 reservoirs individually wrapped; sterile
SDP
Catalog No. NC1706088
Encompass
300 ml Clear Advantage™ reagent reservoirs which are made of crystal clear 100 % virgin polypropylene for improved chemical compatibility. They fit into a reusable automation friendly base with bold, crisp, clearly visible graduation markings.
This item is not returnable. View return policy
Catalog No. NC1706088 Supplier Integra Biosciences Corp Supplier No. 6347
Add to Cart
Add to Cart
300 ml Clear Advantage™ reagent reservoirs which are made of crystal clear 100 % virgin polypropylene for improved chemical compatibility. They fit into a reusable automation friendly base with bold, crisp, clearly visible graduation markings.