Learn More
Ace Glass, Inc. 2000mL reaction kettle complete with sublimation accessories
SDP
Catalog No. 50975894
Encompass_Preferred
2000mL reaction kettle complete with sublimation apparatus accessories. O-ring groove flange on kettle is supplied with a CAPFE o-ring and a quick release clamp. The 300mL condenser flask is connected to the condenser tube with a #25 Ace-Thred nylon bushing. The kettle head features a 34/45 center neck and (3) 24/40 side necks. PTFE sleeves are provided to prevent joint freeze. Also included are (2) 24/40 glass stoppers, and a 24/40 vacuum take-off stopcock adapter.
Catalog No. 50-975-894 Supplier Ace Glass, Inc. Supplier No. 802725
2000mL reaction kettle complete with sublimation apparatus accessories. O-ring groove flange on kettle is supplied with a CAPFE o-ring and a quick release clamp. The 300mL condenser flask is connected to the condenser tube with a #25 Ace-Thred nylon bushing. The kettle head features a 34/45 center neck and (3) 24/40 side necks. PTFE sleeves are provided to prevent joint freeze. Also included are (2) 24/40 glass stoppers, and a 24/40 vacuum take-off stopcock adapter.