Learn More
Ace Glass, Inc. 300ML VESSEL 300ML VESSEL
SDP
Supplier:  Ace Glass, Inc. 669638
Encompass_Preferred
300mL vessel with a 48mm O.D. neck to fit a #50 Ace-Thred, for connection to an umbrella manifold. Vessel has straight sides with stops on the sides to prevent ¿suck in¿ under vacuum. Working capacity of vessel is about 1/2 of volume.
Catalog No. 50-973-325