Learn More
Thermco Products Inc Thermco ACC8540PC Specific Gravity Plain Form Polycarbonate Hydrometers, 1.000 to 1.220 Range, 0.002 Division, 300mm Length
Catalog No. NC0339992
Encompass_Preferred
Thermco’s Plastic Hydrometers are fabricated from polycarbonate (PC) which makes the instrument shatter-proof, while offering glass-like clarity for ease of reading. All models are plain form, so a separate hermometer should be used to measure the temperature of the sample and correct for any temperature variations. Fabricated from PolycarbonateShatter Proof-Glass-Like Clarity-Calibrated against NIST Standards-May be Certified - Traceable to NIST
Catalog No. NC0339992 Supplier Thermco Products Inc Supplier No. ACC8540PC
Thermco’s Plastic Hydrometers are fabricated from polycarbonate (PC) which makes the instrument shatter-proof, while offering glass-like clarity for ease of reading. All models are plain form, so a separate hermometer should be used to measure the temperature of the sample and correct for any temperature variations. Fabricated from PolycarbonateShatter Proof-Glass-Like Clarity-Calibrated against NIST Standards-May be Certified - Traceable to NIST