Learn More
SPI SEMICON 300 mm HWS-RM WaferShipper™
Catalog No. NC1657581
Encompass
The Texchem Advanced Products 300mm HWS-RM Horizontal WaferShiper™ is a unique design providing superior protection and substantial freight cost savings versus competitor HWS and FOSB models. The 300mm HWS-RM is designed for transporting up to 25 thin to full thickness wafers using carbon loaded or non-woven interleaves. By working closely with our material suppliers we develop the cleanest materials for ionics and outgassing, resulting in an ultra clean, carbon filled polypropylene that provides ESD protection.
Catalog No. NC1657581 Supplier SPI SEMICON Supplier No. 300MMHWS_RM
The Texchem Advanced Products 300mm HWS-RM Horizontal WaferShiper™ is a unique design providing superior protection and substantial freight cost savings versus competitor HWS and FOSB models. The 300mm HWS-RM is designed for transporting up to 25 thin to full thickness wafers using carbon loaded or non-woven interleaves. By working closely with our material suppliers we develop the cleanest materials for ionics and outgassing, resulting in an ultra clean, carbon filled polypropylene that provides ESD protection.