Learn More
Chemglass Life Sciences Nest CLS-3499A-024 300 μL Automation Conductive Tips for Hamilton Systems, Sterile, Barcoded, 96 tips x 5 blisters (5,760/case)
SDP
Catalog No. 502733275
Encompass_Preferred
This 300 μL automation tip is intended for high-throughput liquid handling on automated pipetting workstations and provides conductive, barcoded tips supplied pre-sterilized to reduce contamination risk.
  • 300 μL nominal volume per tip.
  • Compatible with Hamilton automated pipetting systems.
  • Conductive medical-grade polypropylene construction for electrostatic compatibility.
  • Pre-sterilized by electron beam; DNase- and RNase-free.
  • Barcoded for automated identification and tracking.
  • Packaged in peelable Tyvek blisters: 96 tips x 5 blisters per pack (480 tips/pack), 5,760 tips per case.
  • Manufactured in a certified cleanroom to minimize contamination.
Catalog No. 50-273-3275 Supplier Chemglass Life Sciences Supplier No. CLS3499A024
This 300 μL automation tip is intended for high-throughput liquid handling on automated pipetting workstations and provides conductive, barcoded tips supplied pre-sterilized to reduce contamination risk.
  • 300 μL nominal volume per tip.
  • Compatible with Hamilton automated pipetting systems.
  • Conductive medical-grade polypropylene construction for electrostatic compatibility.
  • Pre-sterilized by electron beam; DNase- and RNase-free.
  • Barcoded for automated identification and tracking.
  • Packaged in peelable Tyvek blisters: 96 tips x 5 blisters per pack (480 tips/pack), 5,760 tips per case.
  • Manufactured in a certified cleanroom to minimize contamination.