Learn More
Alkali Scientific Bulk Pipet Tips, 300ul, Bagged, 1000/pk
SDP
Catalog No. 502388864
Encompass_Preferred
Bulk Pipet Tips by ASI are ultra-clear and universal fit to most pipettes. Made from 100% polypropylene, low-retention plastic, these tips ensure accurate sample results during liquid handling. The Pipet Tip line is packaged Rnase/Dnase, pyrogen free and available in both sterile and non-sterile bags of 1000 tips.
Catalog No. 50-238-8864 Supplier Alkali Scientific Supplier No. BT1300
Bulk Pipet Tips by ASI are ultra-clear and universal fit to most pipettes. Made from 100% polypropylene, low-retention plastic, these tips ensure accurate sample results during liquid handling. The Pipet Tip line is packaged Rnase/Dnase, pyrogen free and available in both sterile and non-sterile bags of 1000 tips.