Learn More
Alkali Scientific 300uL Hamilton STAR/STARlet Ni
SDP
Catalog No. 503027152
Encompass_Preferred
Alkali Scientificautomatic black conductive 300 L HamiltonSTAR/STARlet Nimbus and Microlab STARplus compatible robotic automation CO-RE style tips are made from high-quality polypropylene (PP) polymer with a surface treatment to enhance hydrophobicity These black conductive tips are precisely engineered for specialized applications offering enhanced efficiency and repeatability in your workflows Material Constructed from medical-grade high-tenacity polypropylene (PP) carefully selected and imported to meet industry standards ensuring consistency and stability between batches Production Environment Manufactured in 100000-class cleanroom facilities to prevent contamination ensuring no DNA RNA protease or endotoxins are present Product point collection and packaging are implemented with traceability maintained through consistent batch numbering enhancing experimental accuracy and minimizing product variation Surface Technology Advanced manufacturing techniques ensure a smooth inner
Catalog No. 50-302-7152 Supplier Alkali Scientific Supplier No. HBFT10300
Alkali Scientificautomatic black conductive 300 L HamiltonSTAR/STARlet Nimbus and Microlab STARplus compatible robotic automation CO-RE style tips are made from high-quality polypropylene (PP) polymer with a surface treatment to enhance hydrophobicity These black conductive tips are precisely engineered for specialized applications offering enhanced efficiency and repeatability in your workflows Material Constructed from medical-grade high-tenacity polypropylene (PP) carefully selected and imported to meet industry standards ensuring consistency and stability between batches Production Environment Manufactured in 100000-class cleanroom facilities to prevent contamination ensuring no DNA RNA protease or endotoxins are present Product point collection and packaging are implemented with traceability maintained through consistent batch numbering enhancing experimental accuracy and minimizing product variation Surface Technology Advanced manufacturing techniques ensure a smooth inner