Learn More
Alkali Scientific 300uL Hamilton STAR/STARlet Ni
SDP
Catalog No. 503027641
Encompass_Preferred
Alkali Scientificautomatic transparent 300 L HamiltonSTAR/STARlet Nimbus and Microlab STARplus compatible robotic automation clear CO-RE style tips are made from high-quality polypropylene (PP) with a special surface treatment that enhances hydrophobicity These transparent tips are precisely engineered for specialized applications offering improved efficiency and repeatability in your processes Material Manufactured from medical-grade high-tenacity polypropylene (PP) carefully selected and imported to meet stringent industry standards ensuring consistent material stability across batches Production Environment Produced in 100000-class cleanroom facilities to ensure the absence of DNA RNA protease and endotoxins The collection and packaging process includes batch numbering for product traceability enhancing experimental precision and reducing variability between products Surface Technology Advanced processing ensures the tips have a smooth inner surface minimizing liquid retention
Catalog No. 50-302-7641 Supplier Alkali Scientific Supplier No. HFT10300
Alkali Scientificautomatic transparent 300 L HamiltonSTAR/STARlet Nimbus and Microlab STARplus compatible robotic automation clear CO-RE style tips are made from high-quality polypropylene (PP) with a special surface treatment that enhances hydrophobicity These transparent tips are precisely engineered for specialized applications offering improved efficiency and repeatability in your processes Material Manufactured from medical-grade high-tenacity polypropylene (PP) carefully selected and imported to meet stringent industry standards ensuring consistent material stability across batches Production Environment Produced in 100000-class cleanroom facilities to ensure the absence of DNA RNA protease and endotoxins The collection and packaging process includes batch numbering for product traceability enhancing experimental precision and reducing variability between products Surface Technology Advanced processing ensures the tips have a smooth inner surface minimizing liquid retention