Learn More
SAI Infusion Technologies 300ul Blood Vials - Sodium Heparinized
SDP
Catalog No. NC0081473
Encompass
300uL Borosilicate glass vials with 100U Sodium Heparin and plastic snap cap, 100/box. Vials are filled and capped in clean room conditions. 6x32mm vials are compatible with some automated blood sampling devices and HPLC systems. Product is for laboratory animal research only.
Catalog No. NC0081473 Supplier SAI Infusion Technologies Supplier No. HV100
300uL Borosilicate glass vials with 100U Sodium Heparin and plastic snap cap, 100/box. Vials are filled and capped in clean room conditions. 6x32mm vials are compatible with some automated blood sampling devices and HPLC systems. Product is for laboratory animal research only.