Learn More
Bellco Glass, Inc. 300 Low Retention Pipet Tip
SDP
Catalog No. 502330241
Encompass_Preferred
CELLTREAT Scientific Products Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing process results in superior low retention. Pipette Tips are universal fit and compatible with major pipettes. Pipette Tips have smooth hydrophobic surfaces to ensure accurate pipetting.Specifications Molded volume reference markings offer a quick double check opportunity Reload System offers a clear window for easy identification of colorcoded wafers Support wafers have large grasping tabs for easy removal from Reload System or Rack Secure locking tab keeps reload system closed and prevents tumbling tips Case Quantity 1920
Catalog No. 50-233-0241 Supplier Bellco Glass, Inc. Supplier No. 229055000
CELLTREAT Scientific Products Pipette Tips deliver high performance with exceptional value. A perfected balance between raw materials and efficient manufacturing process results in superior low retention. Pipette Tips are universal fit and compatible with major pipettes. Pipette Tips have smooth hydrophobic surfaces to ensure accurate pipetting.Specifications Molded volume reference markings offer a quick double check opportunity Reload System offers a clear window for easy identification of colorcoded wafers Support wafers have large grasping tabs for easy removal from Reload System or Rack Secure locking tab keeps reload system closed and prevents tumbling tips Case Quantity 1920