Learn More
Genesee Scientific Corporation Olympus 300 µl Barrier Tips, Filtered Barrier Tip, Low-Binding Resin Technology, Universal Fit, 300 µl
SDP
Catalog No. NC1962415
Encompass
The Olympus 300 µl Barrier Tips are premium filtered pipette tips designed for high-precision liquid handling, featuring low-binding resin technology for maximal sample recovery and contaminant-free certification. Their unique aerosol barrier prevents pipettor contamination, making them ideal for sensitive assays. Universally compatible with most research-grade pipettors, they come in an SBS-format rack for easy identification and ergonomic use.
Catalog No. NC1962415 Supplier Genesee Scientific Corporation Supplier No. 26415
The Olympus 300 µl Barrier Tips are premium filtered pipette tips designed for high-precision liquid handling, featuring low-binding resin technology for maximal sample recovery and contaminant-free certification. Their unique aerosol barrier prevents pipettor contamination, making them ideal for sensitive assays. Universally compatible with most research-grade pipettors, they come in an SBS-format rack for easy identification and ergonomic use.