Learn More
Genesee Scientific Corporation Olympus Plastics™ 300 µL Pipette Tips, Sterile, Low Retention, Ergonomic Design, 300 µL
SDP
Catalog No. NC1909002
Encompass
The Olympus Plastics™ 300 µL Classic Barrier Pipette Tips from Genesee Scientific Corporation enhance liquid handling accuracy with their ergonomic design and low-retention features. These sterile, filtered tips ensure maximum sample recovery while preventing cross-contamination, backed by certifications against common biological contaminants. Their universal fit and racked packaging make them a reliable choice for researchers.
Catalog No. NC1909002 Supplier Genesee Scientific Corporation Supplier No. 24415C
The Olympus Plastics™ 300 µL Classic Barrier Pipette Tips from Genesee Scientific Corporation enhance liquid handling accuracy with their ergonomic design and low-retention features. These sterile, filtered tips ensure maximum sample recovery while preventing cross-contamination, backed by certifications against common biological contaminants. Their universal fit and racked packaging make them a reliable choice for researchers.