Learn More
OCEAN OPTICS INC T300-RT-UV-VIS
Catalog No. NC1767545
Encompass
Transmission dip probes couple to Ocean Insight spectrometers and light sources to measure absorbance and transmission in solutions. These stainless steel probes are great for embedding into process streams for real-time sample monitoring.
Catalog No. NC1767545 Supplier OCEAN OPTICS INC Supplier No. T300RTUVVIS
Transmission dip probes couple to Ocean Insight spectrometers and light sources to measure absorbance and transmission in solutions. These stainless steel probes are great for embedding into process streams for real-time sample monitoring.