Learn More
Medchemexpress LLC 3031336-58-7 | 95.6% | 1151.78 | C58H72ClFN12O8S | 5 MG
SDP
Catalog No. 5000222705
Encompass_Preferred
PROTAC EGFR degrader 2 is a PROTAC designed to selectively degrade the epidermal growth factor receptor (EGFR) and to support biochemical and cellular research into EGFR-driven signaling and antiproliferative effects.
  • Potent EGFR degrader with reported DC50 36.51 nM.
  • Exhibits antiproliferative activity (IC50 ≈4.0 nM).
  • Purity 95.6% suitable for research applications.
  • DMSO solubility up to 100 mg/mL; ultrasonic assistance recommended.
  • Supplied as a light yellow to yellow solid for handling convenience.
  • Store solid at 4°C protected from light; in solution store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-22705 Supplier Medchemexpress LLC Supplier No. HY1443045MG
PROTAC EGFR degrader 2 is a PROTAC designed to selectively degrade the epidermal growth factor receptor (EGFR) and to support biochemical and cellular research into EGFR-driven signaling and antiproliferative effects.
  • Potent EGFR degrader with reported DC50 36.51 nM.
  • Exhibits antiproliferative activity (IC50 ≈4.0 nM).
  • Purity 95.6% suitable for research applications.
  • DMSO solubility up to 100 mg/mL; ultrasonic assistance recommended.
  • Supplied as a light yellow to yellow solid for handling convenience.
  • Store solid at 4°C protected from light; in solution store at -80°C (6 months) or -20°C (1 month).