Learn More
Miele, Inc. A 304 module for phials
Catalog No. NC1469742
Encompass_Preferred
A 304 Module for the optimum loading of up to 98 vials.
Suitable for 98 centrifuge tubes, phials, test tubes, for example
The loading heights differ depending on the positioning
Visit www.mieleusa.com/professional for technical information.
Catalog No. NC1469742 Supplier Miele, Inc. Supplier No. 09862570
A 304 Module for the optimum loading of up to 98 vials.
Suitable for 98 centrifuge tubes, phials, test tubes, for example
The loading heights differ depending on the positioning
Visit www.mieleusa.com/professional for technical information.