Learn More
Biosurface Technologies Corp Stainless Steel (304) Square Coupon, 2 inch x 2 inch
Catalog No. NC0496322
Encompass
This 2 inch by 2 inch square coupon is manufactured from 304 stainless steel, and is designed for biofilm or biofilm-related studies in a laboratory environment. This item has a minimum quantity of 6 per supplier requirements.
Catalog No. NC0496322 Supplier Biosurface Technologies Corp Supplier No. BST 5050304
This 2 inch by 2 inch square coupon is manufactured from 304 stainless steel, and is designed for biofilm or biofilm-related studies in a laboratory environment. This item has a minimum quantity of 6 per supplier requirements.