Learn More
Electron Microscopy Sciences Technovit 3040 Mounting Medium Kit
SDP
Catalog No. 50980430
Encompass_Preferred
Technovit 3040 yellow is a fast curing methyl methacrylate-based resin, whose chemical composition warrants a firm, durable bond between Technovit and the specimen.
Technovit 3040 consists of two components–powder and liquid–allowing simple mixing, easy adherance to the specimen, and fast curing. For fixing the mount, a high viscous consistency is required(i.e. a mixing ratio of approximately 2-3 parts per volume powder: 1 part per volume liquid) has proven to be the most advantageous.
Catalog No. 50-980-430 Supplier Electron Microscopy Sciences Supplier No. 14652
Technovit 3040 yellow is a fast curing methyl methacrylate-based resin, whose chemical composition warrants a firm, durable bond between Technovit and the specimen.
Technovit 3040 consists of two components–powder and liquid–allowing simple mixing, easy adherance to the specimen, and fast curing. For fixing the mount, a high viscous consistency is required(i.e. a mixing ratio of approximately 2-3 parts per volume powder: 1 part per volume liquid) has proven to be the most advantageous.