Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Mechanically Coupled High-Head Centrifugal Pump, 80 GPM/75 ft, 304 SS; 115/230 VAC
Catalog No. NC1688378
Encompass
The mechanically coupled centrifugal pumps, constructed of 304 stainless steel, are ideal for food and pharmaceutical processing plants, as well as water and wastewater treatment centers. They feature high-quality mechanical shaft seals and O-rings. Also available are optional materials for high-temperature and chemical-duty operations. Contact an Application Specialist for more information.
Catalog No. NC1688378 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7072511
The mechanically coupled centrifugal pumps, constructed of 304 stainless steel, are ideal for food and pharmaceutical processing plants, as well as water and wastewater treatment centers. They feature high-quality mechanical shaft seals and O-rings. Also available are optional materials for high-temperature and chemical-duty operations. Contact an Application Specialist for more information.