Learn More
Medchemexpress LLC 306-012B-3 | 00-00-0 | 99.9% | 974.62 g·mol⁻¹ | C46H91N3O6S6 | 5 MG
SDP
Catalog No. 5000221995
Encompass_Preferred
306-O12B-3 is a lipidoid delivery reagent designed for research use to facilitate nucleic acid and small-molecule delivery. It efficiently delivers antisense oligonucleotides (ASO) both in vitro and in vivo and has been used to enhance transport of small molecules across the blood-brain barrier (BBB). The product is supplied as a colorless to light-yellow liquid and is intended for formulation into lipid nanoparticles for experimental applications.
  • Efficient ASO delivery in vitro and in vivo
  • Facilitates transport of small molecules across the blood-brain barrier
  • Ionizable cationic lipidoid suitable for lipid nanoparticle formulations
  • High reported purity (approximately 99.9%)
  • Stable when stored under nitrogen at low temperatures
  • Supplied as a liquid for easy formulation into nanoparticles
Catalog No. 50-002-21995 Supplier Medchemexpress LLC Supplier No. HY1375015MG
306-O12B-3 is a lipidoid delivery reagent designed for research use to facilitate nucleic acid and small-molecule delivery. It efficiently delivers antisense oligonucleotides (ASO) both in vitro and in vivo and has been used to enhance transport of small molecules across the blood-brain barrier (BBB). The product is supplied as a colorless to light-yellow liquid and is intended for formulation into lipid nanoparticles for experimental applications.
  • Efficient ASO delivery in vitro and in vivo
  • Facilitates transport of small molecules across the blood-brain barrier
  • Ionizable cationic lipidoid suitable for lipid nanoparticle formulations
  • High reported purity (approximately 99.9%)
  • Stable when stored under nitrogen at low temperatures
  • Supplied as a liquid for easy formulation into nanoparticles