Learn More
eMolecules 307-24-4 | Perfluorohexanoic acid | Apollo Scientific | MFCD00198040 | 314.054 | C6HF11O2 | 98.000 | OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F | 1g | 562457022
Shop All eMolecules Inc. Products