Learn More
Ace Glass, Inc. 30cm Congealing temperature wire stirrer with #3 stopper
SDP
Catalog No. 50978820
Encompass
30cm congealing temperature wire stirrer with #3 stopper, part of the ACE congealing temperature distillation apparatus, which is similar to that described in U.S. Pharmacopeia, 1995, page 1780. The ACE apparatus is supplied with Ace-Threds in place of rubber stoppers, where practical.
Catalog No. 50-978-820 Supplier Ace Glass, Inc. Supplier No. 654725
30cm congealing temperature wire stirrer with #3 stopper, part of the ACE congealing temperature distillation apparatus, which is similar to that described in U.S. Pharmacopeia, 1995, page 1780. The ACE apparatus is supplied with Ace-Threds in place of rubber stoppers, where practical.