Learn More
SAI Infusion Technologies 30 Gauge, 0.5" Blunt Needles - Non-sterile, 100/pk
SDP
Catalog No. NC9516229
Encompass
30G0.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 100. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.
Catalog No. NC9516229 Supplier SAI Infusion Technologies Supplier No. B3050 100 BULK
30G0.5" Non-Sterile Blunt Needles with plastic hub and stainless steel blunt cannula. These needles are non-sterile, in a bulk case of 100. ETO Exposed sterile blunt needles are available separately. Product is for laboratory animal research only.