Learn More
MSC Justrite Flammable Cabinet, 30 gal, Self-Closing, Yellow, 1 ea
Catalog No. 502886329
Encompass
This self-closing, yellow cabinet is designed for the safe storage of flammable and combustible liquids. It offers a 30-gallon capacity and is built to meet stringent safety standards.
  • Self-closing door mechanism
  • Constructed from durable steel
  • Provides 30 gal capacity for flammable liquids
  • Features two doors and one adjustable shelf
  • Equipped with a 3-point key lock
  • Meets NFPA Code 30 and OSHA standards
  • FM approved
Catalog No. 50-288-6329 Supplier MSC Supplier No. 04858767
This self-closing, yellow cabinet is designed for the safe storage of flammable and combustible liquids. It offers a 30-gallon capacity and is built to meet stringent safety standards.
  • Self-closing door mechanism
  • Constructed from durable steel
  • Provides 30 gal capacity for flammable liquids
  • Features two doors and one adjustable shelf
  • Equipped with a 3-point key lock
  • Meets NFPA Code 30 and OSHA standards
  • FM approved