Learn More
MSC Justrite Standard Cabinet, Manual Closing, 30 gal, Yellow, 1 ea
Catalog No. 502886335
Encompass
This standard storage cabinet is designed for the safe storage of flammable and combustible liquids. It features manual closing doors and a single adjustable shelf, providing a capacity of 30 gal. for vertical storage.
  • Constructed from durable steel
  • Manual closing doors for secure storage
  • Designed for flammable and combustible liquids
  • Equipped with adjustable shelves
  • Keyed lock with 3-point key
  • Yellow finish
  • Two doors and one shelf
  • Meets NFPA Code 30, OSHA, and NFPA standards
  • FM Approved
Catalog No. 50-288-6335 Supplier MSC Supplier No. 75340091
This standard storage cabinet is designed for the safe storage of flammable and combustible liquids. It features manual closing doors and a single adjustable shelf, providing a capacity of 30 gal. for vertical storage.
  • Constructed from durable steel
  • Manual closing doors for secure storage
  • Designed for flammable and combustible liquids
  • Equipped with adjustable shelves
  • Keyed lock with 3-point key
  • Yellow finish
  • Two doors and one shelf
  • Meets NFPA Code 30, OSHA, and NFPA standards
  • FM Approved