Learn More
Felix Storch Inc SUMMIT 30IN WIDE TOP FREEZER/REFRIGER
SDP
Catalog No. NC3343324
Encompass_Preferred
The CTR21PL is a full-sized refrigerator-freezer. The large interior offers 20.5 cu.ft. of storage capacity, with frost-free operation for low maintenance use. It comes in a textured gray cabinet with user-reversible fingerprint-free doors that offer the look of stainless steel without the constant cleanup. Left hand door swing: https://www.accucold.com/product/CTR21PLLHD
Catalog No. NC3343324 Supplier Felix Storch Inc Supplier No. CTR21PLLHD
The CTR21PL is a full-sized refrigerator-freezer. The large interior offers 20.5 cu.ft. of storage capacity, with frost-free operation for low maintenance use. It comes in a textured gray cabinet with user-reversible fingerprint-free doors that offer the look of stainless steel without the constant cleanup. Left hand door swing: https://www.accucold.com/product/CTR21PLLHD