Learn More
Ace Glass, Inc. Mantle, 30L, for 276.9mm diameter cylindrical flask, 276.9mm ID x 79.2mm height, 370w-115v, bottom opening, .
SDP
Catalog No. 50977334
Encompass
30L aluminum housed, table-model mantle with bottom opening for 276.9mm diameter cylindrical flask. 276.9mm I.D. x 79.2mm height. Maximum internal operating temperature is 450?C. 370w-115v, and is cULus rated. Power cord included.
Catalog No. 50-977-334 Supplier Ace Glass, Inc. Supplier No. 1205354
30L aluminum housed, table-model mantle with bottom opening for 276.9mm diameter cylindrical flask. 276.9mm I.D. x 79.2mm height. Maximum internal operating temperature is 450?C. 370w-115v, and is cULus rated. Power cord included.