Learn More
Ace Glass, Inc. Mantle, 30L, for 317.5mm diameter cylindrical flask, 276.9mm ID x 79.3mm height, 370w-115v, CSA rated
SDP
Catalog No. 50977333
Encompass
30L aluminum housed, table-model mantle for 317.5mm diameter cylindrical flask. 276.9mm I.D. x 79.3mm height. Maximum internal operating temperature is 450 Deg. centigrade. 370w-115v, cULus rated. Power cord included.
Catalog No. 50-977-333 Supplier Ace Glass, Inc. Supplier No. 1205345
30L aluminum housed, table-model mantle for 317.5mm diameter cylindrical flask. 276.9mm I.D. x 79.3mm height. Maximum internal operating temperature is 450 Deg. centigrade. 370w-115v, cULus rated. Power cord included.