Learn More
Alkali Scientific 30L Automation Tips P30 for Ag
SDP
Supplier:  Alkali Scientific ABFT1030
Encompass_Preferred
Alkali ScientificP30 automation tips are designed for optimal compatibility with Agilent Bravo and Agilent VPrep robotic liquid handlers These tips eliminate the need for a separate labware definition ensuring seamless integration The Agilent Bravo is a flexible liquid handling platform that automates your sample preparation for screening applications such as compound management cell-based assays and genomic assays With this robotic liquid handling system time is freed up and consistent data are achieved across samples and users The compact liquid handler can be easily placed on a small bench and fits in a flow hood With more than 60 accessories ranging from thermal cycler to shaker the system can be tailored to automate most sample preparation protocols Automating your sample prep protocol has never been easier with the included VWorks control software that allows users to create and run flexible liquid handling protocols quickly Specifications Working volume 0 25 30 00 L 0
Catalog No. 50-302-6435