Learn More
Chemglass Life Sciences 30L Process Reactor, Complete, Electric Motor, 82.6" OAH with Motor, 20.9" Clearance Below Valve
SDP
Catalog No. CG1972630
Encompass_Preferred
30L Process Reactor, Complete, Electric Motor, 82.6" OAH with Motor, 20.9" Clearance Below Valve. Complete reactor system on our NEW open air stand.Features:¿ Motor Mount Arm Can Unlock And Swing Out¿ Smaller Footprint Support Frame With Oversized Rugged S.S. Uprights And Heavy Duty PTFE Coated Aluminum Fittings¿ Electric And XP Motor Has Separate Controller With Digital Display¿ Reactor Support Plate Holds Reactor Securely And Allows Easy Installation/Removal Of Reactor By One Person¿ Lower Safety Ring And Lower Mantle Base¿ Heavy Duty Lockable Casters ¿ Electric Or Air Motor Optional XP Motor Available¿ Open Frame Design Provides Excellent Accessibility¿ 7-Neck Lid With Large Powder Addition Port¿ Ridge Clamp Accepts 1/2¿ Rod And Adds Additional Support For Large Glassware¿ #304 S.S. Cooling Coil PTFE Coated ¿ Digital Temperature Controller¿ Zero Dead Space Detachable Drain Valve Has Knob With Laser Etched Directional Arrows For Open And Close
Catalog No. CG1972630 Supplier Chemglass Life Sciences Supplier No. CG1972630
Add to Cart
Add to Cart
Request a Quote
30L Process Reactor, Complete, Electric Motor, 82.6" OAH with Motor, 20.9" Clearance Below Valve. Complete reactor system on our NEW open air stand.Features:¿ Motor Mount Arm Can Unlock And Swing Out¿ Smaller Footprint Support Frame With Oversized Rugged S.S. Uprights And Heavy Duty PTFE Coated Aluminum Fittings¿ Electric And XP Motor Has Separate Controller With Digital Display¿ Reactor Support Plate Holds Reactor Securely And Allows Easy Installation/Removal Of Reactor By One Person¿ Lower Safety Ring And Lower Mantle Base¿ Heavy Duty Lockable Casters ¿ Electric Or Air Motor Optional XP Motor Available¿ Open Frame Design Provides Excellent Accessibility¿ 7-Neck Lid With Large Powder Addition Port¿ Ridge Clamp Accepts 1/2¿ Rod And Adds Additional Support For Large Glassware¿ #304 S.S. Cooling Coil PTFE Coated ¿ Digital Temperature Controller¿ Zero Dead Space Detachable Drain Valve Has Knob With Laser Etched Directional Arrows For Open And Close