Learn More
Chemglass Life Sciences Capacity Liters: 30, Flange I.D. (mm): 300, Valve Style: 2" BP Detachable, Overall Height mm (in.): 530 (20.9), Height to Width Ratio: 1.6:1
SDP
Catalog No. CG197261
Encompass_Preferred
Reaction vessels, unjacketed, Capacity Liters: 30, Flange I.D. (mm): 300, Valve Style: 2" BP Detachable, Overall Height mm (in.): 530 (20.9), Height to Width Ratio: 1.6:1.Unjacketed large scale process reaction vessels. The 10, 15, and 20L vessels have a 200mm ID flange with an o-ring groove formed in the surface of the flange. The 30L and 50L have a 300mm ID flat ground flange. The 10, 15, and 20L are available with a choice of a 2¿ beaded pipe drain that is detachable, or the 0-20mm valve with 3/4¿ sidearm that is permanently sealed. These flush bottom (zero dead space) valves are designed to prevent solids from collecting in the bottom of the vessel. Vessels equipped with the 0-20mm sealed on valve are supplied complete with a TFE plug having Perfluoro chemically resistant o-rings. Vessels are serial numbered and have a graduation scale on both sides showing the approximate volume.PLEASE NOTE: VALVE, BEADED PIPE COUPLINGS, AND ADAPTERS MUST BE PURCHASED SEPARATELY.
Catalog No. CG197261 Supplier Chemglass Life Sciences Supplier No. CG197261
Reaction vessels, unjacketed, Capacity Liters: 30, Flange I.D. (mm): 300, Valve Style: 2" BP Detachable, Overall Height mm (in.): 530 (20.9), Height to Width Ratio: 1.6:1.Unjacketed large scale process reaction vessels. The 10, 15, and 20L vessels have a 200mm ID flange with an o-ring groove formed in the surface of the flange. The 30L and 50L have a 300mm ID flat ground flange. The 10, 15, and 20L are available with a choice of a 2¿ beaded pipe drain that is detachable, or the 0-20mm valve with 3/4¿ sidearm that is permanently sealed. These flush bottom (zero dead space) valves are designed to prevent solids from collecting in the bottom of the vessel. Vessels equipped with the 0-20mm sealed on valve are supplied complete with a TFE plug having Perfluoro chemically resistant o-rings. Vessels are serial numbered and have a graduation scale on both sides showing the approximate volume.PLEASE NOTE: VALVE, BEADED PIPE COUPLINGS, AND ADAPTERS MUST BE PURCHASED SEPARATELY.