Learn More
Chemglass Life Sciences Reactor Support Frame, 30L with 13.8" Valve Clearance OR 50L (Short) with 10.5" Valve Clearance, 28.25" W x 20" D x 66.50" H
SDP
Catalog No. 501531339
Encompass_Preferred
Reactor Support Frame, 30L with 13.8" Valve Clearance OR 50L (Short) with 10.5" Valve Clearance, 28.25" W x 20" D x 66.50" H. Open air design, with decreased frame depth, allows greater access to the reactor and peripheral glassware. Motor mount arm can be used with either electric, air, or XP motors and can swing out of the way for greater access to reactor. Design allows support frame to be used for 30L through 50L Jacketed or Unjacketed reactor systems. Glassware can be quickly installed and removed by one person saving time and frustration normally associated with out-of-date designs.
Catalog No. 50-153-1339 Supplier Chemglass Life Sciences Supplier No. CG1968X10
Add to Cart
Add to Cart
Request a Quote
Reactor Support Frame, 30L with 13.8" Valve Clearance OR 50L (Short) with 10.5" Valve Clearance, 28.25" W x 20" D x 66.50" H. Open air design, with decreased frame depth, allows greater access to the reactor and peripheral glassware. Motor mount arm can be used with either electric, air, or XP motors and can swing out of the way for greater access to reactor. Design allows support frame to be used for 30L through 50L Jacketed or Unjacketed reactor systems. Glassware can be quickly installed and removed by one person saving time and frustration normally associated with out-of-date designs.